Learn More
Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, 98%, BLD Pharm™
Product Code. 30972827
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30972827 500 g
30969441 100 g
30970802 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30972827 Supplier BLD Pharm Supplier No. BD106052500g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 4468-02-4
Molecular Formula C12H22O14Zn
MDL Number MFCD00868110
Quantity 500 g
SMILES O=C([O-])[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.O=C([O-])[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.[Zn+2]
Molecular Weight (g/mol) 455.70
Percent Purity 98%
Chemical Name or Material Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate