missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059255
915.19 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]

Zinc oxide is incorporated in powders, ointments, and pastes. As Accelerator activator, pigment and reinforcing agent in rubber, ointments, pigment and mould-growth inhibitor in paints, uv absorber in plastics, ceramics, floor tile, glass, zinc salts, feed additive, dietary supplement, seed treatment cosmetics, photoconductor in office copying machines and in color photography, piezoelectric devices, artists' colorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1975°C
Physical Form Tablets
Quantity 25 g
UN Number UN3077
Merck Index 14,10147
Solubility Information Insoluble in water.
Formula Weight 81.37
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc oxide, 10 to 12mm dia. x 4 to 5mm thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'