missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Catalog Number p-7073671
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
Melting Point 1975°C
Color White
pH 7
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 81.38g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Zinc oxyde
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'