Läs mer
Zinc Pentamethylenedithiocarbamate, TRC
Produktkod. 30455860
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30455860 100 mg
1 options
Produktkod. 30455860 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ701665100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Zinc Pentamethylenedithiocarbamate
CAS 13878-54-1
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H20 N2 S4 Zn
Kvantitet 100 mg
Synonym Zinc','bis(1-piperidinecarbodithioato-κS1,κS1')-','(T-4)-, Zinc','bis(1-piperidinecarbodithioato)- (8CI), Zinc','bis(1-piperidinecarbodithioato-S,S')-','(T-4)-, 1-Piperidinecarbodithioic acid','zinc complex, Accelerator ZPD, Bis(piperidinocarbodithioato)zinc, Nocceler ZP, Vulkacit ZP, Vulkacit ZPD, ZP, Zinc N,N-pentamethylenedithiocarbamate, Zinc pentamethylenedithiocarbamate
InChI formel InChI=1S/2C6H11NS2.Zn/c2*8-6(9)7-4-2-1-3-5-7;/h2*1-5H2,(H,8,9);/q;;+2/p-2
LEDER C1CCN(CC1)C(=S)[S-].C1CCN(CC1)C(=S)[S-].[Zn+2]
Molekylvikt (g/mol) 385.94
Formel vikt 383.9801