missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc selenide, Thermo Scientific Chemicals
233.53 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
Specifications
Melting Point | >1100°C |
Quantity | 10 g |
UN Number | UN3283 |
Merck Index | 14,10156 |
Solubility Information | Insoluble in water. |
Formula Weight | 144.33 |
Odor | Odorless |
Density | 5.42^1^5 |
Chemical Name or Material | Zinc selenide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'