missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051782
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1315-09-9
SeZn
144.35
MFCD00011296
GTLQJUQHDTWYJC-UHFFFAOYSA-N
zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
zinc(2+) selandiide
[Zn++].[Se--]
This item is not returnable. View return policy
1315-09-9
SeZn
144.35
MFCD00011296
GTLQJUQHDTWYJC-UHFFFAOYSA-N
zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
zinc(2+) selandiide
[Zn++].[Se--]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >1100°C
Physical Form Powder
Quantity 25 g
UN Number UN3283
Merck Index 14,10156
Solubility Information Insoluble in water; Decomposes in dilute HNO3
Formula Weight 144.33
Percent Purity 99.99%
Odor Odorless
Sensitivity Air Sensitive
Density 5.42 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Zinc selenide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'