Läs mer
Palintest™ Zinc Starter Tdisc Test
Produktkod. 11998896
Kvantitet:
50-pack
Produktkod. 11998896
Brand: Palintest™ CM148
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
For use with Palintest™ comparator
The Palintest™ comparator with interchangeable colour discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifikationer
Koncentration 0 to 4 mg/L
Kvantitet 50-pack