Learn More
Zinc trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Catalog Number p-3756745
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
54010-75-2
C2F6O6S2Zn
363.51
MFCD00013229
CITILBVTAYEWKR-UHFFFAOYSA-L
zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
104671
zinc;trifluoromethanesulfonate
[Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
54010-75-2
C2F6O6S2Zn
363.51
MFCD00013229
CITILBVTAYEWKR-UHFFFAOYSA-L
zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
104671
zinc;trifluoromethanesulfonate
[Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Gray to White
Physical Form Powder
Quantity 10 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CF3SO3)2Zn
Solubility Information Solubility in water: very soluble
Formula Weight 363.51
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Zinc trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Zinc trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.