Läs mer
Zineb, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Zineb |
| CAS | 12122-67-7 |
| Molekylformel | C4 H6 N2 S4 Zn |
| Kvantitet | 5 g |
| Synonym | Zinc','[N-[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, Zinc','[[1,2-ethanediylbis[carbamodithioato]](2-)]-, Zinc','[[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']- (9CI), Zinc','[ethylenebis[dithiocarbamato]]- (8CI), Carbamodithioic acid','1,2-ethanediylbis-','zinc complex, Aaphytora, Aphytora, Aspor, Aspor C, Asporum, Azzurro, Blizene, Bonzeb, Bonzeb 10, Carbadine, Carbamodithioic acid','1,2-ethanediylbis-','zinc salt, Crittox, Curit, Cynkotox, Daisen, Deikusol, Dipher, Discon, Dithane 65, Dithane Z, Dithane Z 78, Ethylenebis[dithiocarbamato]zinc, Ethylenebis[dithiocarbamic acid] zinc salt, Fungo-Pulvit, Funjeb, Hexathane, Indofil Z 78, Lirotan, Lonacol, Micide 55, NSC 49513, Novozir, Novozir N, Novozir N 50, Parzate zineb, Perozin, Perozin 75B, Perozine, Perozine 75B, Phytox, Pilzol SZ, Polyram Z, Thionic M, Tiezene, Tritoftorol, Turbair Zineb Fungicide, Unizeb, Zebenide, Zebtox, Zinc N,N'-ethylenebisdithiocarbamate, Zinc ethylenebis[dithiocarbamate], Zineb, Zineb 80, Zinosan |
| InChI formel | InChI=1S/C4H8N2S4.Zn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2 |
| LEDER | S=C1NCCNC(=S)S[Zn]S1 |
| IUPAC-namn | 1,3-dithia-5,8-diaza-2λ^{2}-zincacyclononane-4,9-dithione |
| Molekylvikt (g/mol) | 275.77 |
| Formel vikt | 273.8705 |