Läs mer
Zinquin Ethyl Ester, TRC
Produktkod. 30455689
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455689 5 mg
30457219 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ4500005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 181530-09-6
Molekylformel C21 H22 N2 O5 S
Molekylvikt (g/mol) 414.47
Synonym 2-[[2-Methyl-8-[[(4-methylphenyl)sulfonyl]amino]-6-quinolinyl]oxy]acetic Acid, Zinquin Zinquin A, [[2-Methyl-8-[[(4-methylphenyl)sulfonyl]amino]-6-quinolinyl]oxy]-acetic Acid
IUPAC-namn ethyl 2-[2-methyl-8-[(4-methylphenyl)sulfonylamino]quinolin-6-yl]oxyacetate
LEDER CCOC(=O)COc1cc(NS(=O)(=O)c2ccc(C)cc2)c3nc(C)ccc3c1
Specifikationer
Kemiskt namn eller material Zinquin Ethyl Ester
Kvantitet 5 mg
InChI formel InChI=1S/C21H22N2O5S/c1-4-27-20(24)13-28-17-11-16-8-7-15(3)22-21(16)19(12-17)23-29(25,26)18-9-5-14(2)6-10-18/h5-12,23H,4,13H2,1-3H3
Formel vikt 414.1249