Läs mer
Emma Safety Footwear Zion XD Safety Shoes
Produktkod. 15978301
Change view
Storlek:
39
40
41
42
43
44
45
46
47
48
49
Förpackningsstorlek:
1 par
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Storlek unitSize
15978301 43 1 par
15938301 39 1 par
15948301 40 1 par
15958301 41 1 par
15968301 42 1 par
15988301 44 1 par
15998301 45 1 par
15908311 46 1 par
15918311 47 1 par
15928311 48 1 par
15938311 49 1 par
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Produktkod. 15978301 Leverantör Emma Safety Footwear Leverantörsnummer MM30186843
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical resistant with highly visible 3M™ reflective strip. The impenetrable sole protects feet from sharp and metal objects while the steel toe cap and reinforced polyurethane toe cover offer protection from falling objects. Microfiber components stand up to cement and remains flexible.
  • S3-Safety shoe with sole plate for wet work
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Design Zion XD
Material Microfiber
Storlek 43