Läs mer
Ziprasidone hydrochloride monohydrate, MedChemExpress
Produktkod. 16483337
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16483337 50 mg
16437328 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16483337 Leverantör Medchem Express Leverantörsnummer HY17407
MedChemExpress Ziprasidone (CP-88059) hydrochloride monohydrate is an orally active combined 5-HT and dopamine receptor antagonist. Ziprasidone hydrochloride monohydrate has affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM) and 5-HT1A (Ki=3.4 nM).
Ziprasidone (CP-88059) hydrochloride monohydrate is an orally active combined 5-HT and dopamine receptor antagonist. Ziprasidone hydrochloride monohydrate has affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM) and 5-HT1A (Ki=3.4 nM).
Specifikationer
Kemiskt namn eller material Ziprasidone hydrochloride monohydrate
Fysisk form Solid
CAS 138982-67-9
Färg Pink
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H24Cl2N4O2S
För användning med (applikation) Neuroscience-Neuromodulation
Synonym CP 88059 hydrochloride monohydrate
Löslighetsinformation DMSO : 25 mg/mL (53.49 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.14 mM; Need ultrasonic and warming)
LEDER O=C1CC2=CC(CCN3CCN(C4=NSC5=C4C=CC=C5)CC3)=C(Cl)C=C2N1.[H]Cl.O
Molekylvikt (g/mol) 467.41
Formel vikt 467.41
Procent renhet 99.74%