missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
Description
Zirconium Boride is used as an aerospace refractory, in cutting tools, and to protect thermocouple tubes. It is also acts as an antioxidant in carbon-bonded refractory ceramics, neutron absorber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 3000°C |
Physical Form | Powder |
Quantity | 250 g |
Linear Formula | ZrB2 |
UN Number | UN3178 |
Solubility Information | It is insoluble in water. |
Formula Weight | 112.84 |
Percent Purity | 99.5% |
Assay | (metals basis excluding Hf) |
Chemical Name or Material | Zirconium boride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'