missing translation for 'onlineSavingsMsg'
Learn More

Chempur ZIRCONIUM FOIL 0.25 X 150 X 150 MM 99.8%,

Brand:  Chempur x.901096

Encompass

ZIRCONIUM FOIL 0.25 X 150 X 150 MM 99.8%,

Product Code. 11630212

  • 11135.00 SEK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy