Learn More
S100A13 Antibody (020), PerCP, R&D Systems™
Product Code. 18614831 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18614831 0.1 mL 0.1mL
1 options
Product Code. 18614831 Supplier Novus Biologicals Supplier No. NBP289734PCP
 Restricted Item
Beräknad leverans: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
S100A13 Monoclonal specifically detects S100A13 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antigen S100A13
Applications ELISA
Classification Monoclonal
Clone 020
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A13 (Accession#: NP_001019381.1; Met1-Lys98).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6284
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus