Läs mer
Thermo Scientific™ Zirconium Oxide Round Bottom Crucibles, Low Form
Kapacitet (metrisk):
5 mL
100 mL
200 mL
Diameter (metrisk) Yttre:
29mm
67mm
82mm
Höjd (metrisk):
21mm
51mm
63mm
Förpackningsstorlek:
Varje
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
Specifikationer
| Material | Zirconium Oxide |
| Kvantitet | 1 Piece |
| Kapacitet (metrisk) | 5 mL |
| Diameter (metrisk) Yttre | 29mm |
| Höjd (metrisk) | 21mm |
| Produkttyp | Zirconium Oxide Round Bottom Crucibles, Low Form |
| Diameter (metrisk) bas | 18mm |