Läs mer
Tocris Bioscience™ ZM 241385
GREENER_CHOICE
Produktkod. 30001169
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001169 50 mg
30000702 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001169 Leverantör Tocris Bioscience™ Leverantörsnummer 1036/50
Potent, highly selective A2A antagonist
Potent and highly selective A2A adenosine antagonist, with a pA2 of 9.02 for A2A receptors in guinea pig cardiac vasculature and fold selectivities of 1000, 91 and 500,000 over A1, A2B and A3 sites respectively. Active in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139180-30-6
Molekylformel C16H15N7O2
Specifikationer
Kemiskt namn eller material ZM 241385
Kvantitet 50 mg
Formel vikt 337.34
Renhet >99%
Molekylvikt (g/mol) 337.343