Läs mer
Tocris Bioscience™ ZM 323881 hydrochloride
GREENER_CHOICE
Produktkod. 18095262
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18095262 1 mg
18085682 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18095262 Leverantör Tocris Bioscience™ Leverantörsnummer 2475/1
Potent, selective inhibitor of VEGFR-2
Potent and selective inhibitor of human vascular endothelial growth factor receptor 2 (VEGFR-2/KDR) activity. Selectively inhibits VEGFR-2 (IC50 = 2 nM) over VEGFR-1 and a range of other receptor tyrosine kinases such as PDGFRβ, FGFR1, EGFR and erbB2 (IC50 > 50 μM). Inhibits VEGF-A-induced endothelial cell proliferation in vitro (IC50 = 8 nM).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 193000-39-4
Molekylformel C22H19ClFN3O2
Specifikationer
Kemiskt namn eller material ZM 323881 hydrochloride
Kvantitet 1 mg
Formel vikt 411.86
Renhet >99%
Molekylvikt (g/mol) 411.861