missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF251 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ZNF251 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884165
|
Proteintech
25601-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874165
|
Proteintech
25601-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZNF251 may be involved in transcriptional regulation.Specifications
ZNF251 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9BRH9 | |
71591, 90987 | |
ZNF251 Fusion Protein Ag22347 | |
Primary | |
-20°C | |
ZNF251 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
zinc finger protein 251, ZNF251 | |
Zfp251, ZNF251 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |