missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF266 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11374-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing many tandem zinc-finger motifs. Zinc fingers are protein or nucleic acid-binding domains, and may be involved in a variety of functions, including regulation of transcription. This gene is located in a cluster of similar genes encoding zinc finger proteins on chromosome 19. Alternative splicing results in multiple transcript variants for this gene.Specifications
ZNF266 | |
Polyclonal | |
Unconjugated | |
ZNF266 | |
HZF1, KIAA2007, zinc finger protein 266, Zinc finger protein HZF1, ZNF266 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10781 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14584 | |
ZNF266 | |
ZNF266 Fusion Protein Ag1933 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |