missing translation for 'onlineSavingsMsg'
Learn More

ZNF266 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11374-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878403

  • 4721.67 SEK / 150µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein containing many tandem zinc-finger motifs. Zinc fingers are protein or nucleic acid-binding domains, and may be involved in a variety of functions, including regulation of transcription. This gene is located in a cluster of similar genes encoding zinc finger proteins on chromosome 19. Alternative splicing results in multiple transcript variants for this gene.
Specifications

Specifications

ZNF266
Polyclonal
Unconjugated
ZNF266
HZF1, KIAA2007, zinc finger protein 266, Zinc finger protein HZF1, ZNF266
Rabbit
Antigen Affinity Chromatography
RUO
10781
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14584
ZNF266
ZNF266 Fusion Protein Ag1933
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers