Learn More
ZNF287 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22162-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.Specifications
ZNF287 | |
Polyclonal | |
Unconjugated | |
ZNF287 | |
zinc finger protein 287, ZKSCAN13, ZNF287 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57336 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HBT7 | |
ZNF287 | |
ZNF287 Fusion Protein Ag17814 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |