missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ZNF514 (Human) Recombinant Protein
Human ZNF514 full-length ORF ( NP_116177.1, 1 a.a. - 400 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00084874-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTFEDVAVEFSQWEWGQLNPAQKDLYREVMLENFRNLAILGLLVSKPYVICQLEEGGEPFMVEREISTGAHSDWKRRSKSKESMPSWGISKEELFQVVSVEKHIQDVLQFSKLKAACGCDGQLEMQQIKQERHLKQMSTIHKSATTLSRDYKWNGFGRSLGLRSVLVNQHSILMGEGSYKCDTEFRQTLGGNNSQRTHPEKKSCKCNECGKSFHFQSELRRHQRCHTGEKPYECSDCGRAFGHISSLIKHQRTHTGEKPYECSECGRAFSQSSSLVLHYRFHTGEKPYKCNECGRAFGHTSSLIKHQRTHTGEKPYECRECGRTFSQSSSLIVHYRFHTGEKPYKCNKCGRAFSQSSSLTQHYRFHTGEKPYKCNECGRAFAHTASLIKHQRSHAGKKTL
Specifications
NP_116177.1 | |
zinc finger protein 514 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZNF514 | |
GST |
84874 | |
Wheat germ expression system | |
10 μg | |
MGC126229, MGC126230 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |