missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF597 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20922-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May be involved in transcriptional regulation.Specifications
ZNF597 | |
Polyclonal | |
Unconjugated | |
ZNF597 | |
zinc finger protein 597, ZNF597 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146434 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96LX8 | |
ZNF597 | |
ZNF597 Fusion Protein Ag15183 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |