Läs mer
Invitrogen™ ZO-1 Monoclonal Antibody (R26.4C), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
16850.00 SEK - 38090.00 SEK
Specifikationer
| Antigen | ZO-1 |
|---|---|
| Klona | R26.4C |
| Koncentration | 0.5 mg/mL |
| Klassificering | Monoclonal |
| Konjugera | Unconjugated |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
17140782
|
Invitrogen™
14-9776-37 |
2 mg |
16850.00 SEK
2mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17150782
|
Affymetrix eBioscience
14-9776-95 |
10 mg | |||||||
Beskrivning
The MAb R26.4C recognizes the tight junction protein 1, ZO-1 (zona occludens) of rat, mouse, canine, and porcine. ZO-1 is a member of the MAGUK family of tight junction associated scaffolding proteins, each containing a PDZ, SH3 and guanylate kinase-like (GUK) domain. ZO-1 is an integral member of the tight junction complex which forms a barrier to paracellular movement of substances, separating apical and basolateral fluids in relation to the epithelial cell layer. ZO-1 forms complexes with family members ZO-2, ZO-3, as well as occludin, cingulin, claudin-1-8, actin, alpha-catenin, the Ras-effector molecule AF-6, ZONAB, Apg-2, and the junctional adhesion molecules (JAM)-A-C. ZO-1 is thought to anchor the actin cytoskeleton to the tight junction. ZO-1 and ZO-2 have been shown to regulate cell cycle and proliferation with recent evidence highlighting the down-regulation of ZO-1 and ZO-2 with cancer progression.
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.Specifikationer
| ZO-1 | |
| 0.5 mg/mL | |
| Unconjugated | |
| Rat | |
| Mouse, Rat, Canine | |
| O97758, P39447 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| R26.4C | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 21872, 292994, 403752 | |
| Primary | |
| 4°C | |
| TJP1 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.