Learn More
ZO-2 Mouse anti-Human, Unconjugated, Clone: 1D2E5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67387-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839985
  • 1656.73 SEK / 20µL
Estimated Shipment: 23-04-2024
to see stock.
Description
Description
The ZO-2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZO-2. This antibody recognizes human, rat antigen. The ZO-2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ZO-2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, ELISA
1D2E5
Unconjugated
20 μL
9414
IgG2a