Learn More
ZO-2 Mouse anti-Human, Unconjugated, Clone: 1D2E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67387-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The ZO-2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZO-2. This antibody recognizes human, rat antigen. The ZO-2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ZO-2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Western Blot, ELISA | |
1D2E5 | |
Unconjugated | |
20 μL | |
9414 | |
IgG2a |