Läs mer
Zoligratinib, MedChemExpress
Produktkod. 16483716
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16483716 25 mg 25mg
16473907 50 mg -
16450448 5 mg 5mg
16407988 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16483716 Leverantör Medchem Express Leverantörsnummer HY19957
MedChemExpress Zoligratinib (Debio 1347) is an orally available and selective FGFR inhibitor with IC50s of 9.3, 7.6, and 22 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.
Zoligratinib (Debio 1347) is an orally available and selective FGFR inhibitor with IC50s of 9.3, 7.6, and 22 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.
Specifikationer
Kemiskt namn eller material Zoligratinib
Fysisk form Solid
CAS 1265229-25-1
Färg Light Yellow
Kvantitet 25 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H16N6O
För användning med (applikation) Cancer-Kinase/protease
Synonym Debio 1347 CH5183284
Löslighetsinformation DMSO : 25 mg/mL (70.15 mM; ultrasonic and warming and heat to 50°C)
LEDER O=C(C1=C(N)N(C2=CC=C3N=C(C)NC3=C2)N=C1)C(N4)=CC5=C4C=CC=C5
Molekylvikt (g/mol) 356.38
Formel vikt 356.38
Procent renhet 99.32%