Läs mer
Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde), Mikromol™
Produktkod. 17952216
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17952216 100 mg
17944176 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17952216 Leverantör Mikromol Leverantörsnummer MM0333.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde)
CAS 400777-11-9
Analyt- eller komponentnamn Zolpaldehyde (6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde)
Frakt skick Ice pack (-20°C)
Molekylformel C16 H14 N2 O
Certifikat och efterlevnad ISO 17025
Synonym Zolpaldehyde, 6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carboxaldehyde
InChI formel InChI=1S/C16H14N2O/c1-11-3-6-13(7-4-11)16-14(10-19)18-9-12(2)5-8-15(18)17-16/h3-10H,1-2H3
LEDER Cc1ccc(cc1)c2nc3ccc(C)cn3c2C=O
IUPAC-namn 6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-carbaldehyde
Molekylvikt (g/mol) 250.3
Kvantitet 100 mg
Formel vikt 250.1106 g/mol