Läs mer
Zolpidem 6-Carboxylic Acid Methyl Ester, TRC
Produktkod. 30453928
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30453928 5 mg
1 options
Produktkod. 30453928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ6500255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Zolpidem 6-Carboxylic Acid Methyl Ester
CAS 917252-81-4
Molekylformel C20H21N3O3
Kvantitet 5 mg
Synonym 3-[2-(Dimethylamino)-2-oxoethyl]-2-(4-methylphenyl)-imidazo[1,2-a]pyridine-6-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C20H21N3O3/c1-13-5-7-14(8-6-13)19-16(11-18(24)22(2)3)23-12-15(20(25)26-4)9-10-17(23)21-19/h5-10,12H,11H2,1-4H3
LEDER CC1=CC=C(C=C1)C2=C(N3C=C(C=CC3=N2)C(=O)OC)CC(=O)N(C)C
IUPAC-namn methyl 3-[2-(dimethylamino)-2-oxoethyl]-2-(4-methylphenyl)imidazo[1,2-a]pyridine-6-carboxylate
Molekylvikt (g/mol) 351.4
Formel vikt 351.1583