Läs mer
Zomepirac Acyl-O-Beta-D-glucuronide, TRC
Produktkod. 30450285
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30450285 2.5 mg 2.5mg
30453361 1 mg -
30454203 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ6750102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75871-31-7
Molekylformel C21H22ClNO9
Molekylvikt (g/mol) 467.85
Synonym 1-[5-(4-Chlorobenzoyl)-1,4-dimethyl-1H-pyrrole-2-acetate] β-D-Glucopyranuronic Acid, Zomepirac Glucuronide, Zomepirac Acyl-1-O-β-D-glucuronide
LEDER CC1=C(N(C(=C1)CC(=O)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O)C)C(=O)C3=CC=C(C=C3)Cl
Specifikationer
Kemiskt namn eller material Zomepirac Acyl-O-beta-D-glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C21H22ClNO9/c1-9-7-12(23(2)14(9)15(25)10-3-5-11(22)6-4-10)8-13(24)31-21-18(28)16(26)17(27)19(32-21)20(29)30/h3-7,16-19,21,26-28H,8H2,1-2H3,(H,29,30)/t16-,17-,18+,19-,21+/m0/s1
Formel vikt 467.0983
Procent renhet >95%