Läs mer
Zopiclone N-Oxide, TRC
Produktkod. 30008557
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008557 50 mg
30012008 5 mg
30008282 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ70051050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43200-96-0
Molekylformel C17 H17 Cl N6 O4
Molekylvikt (g/mol) 404.81
Synonym 1-Piperazinecarboxylic acid, 4-methyl-, 6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, 4-oxide, 5H-Pyrrolo[3,4-b]pyrazine, 1-piperazinecarboxylic acid deriv., RP 29753, Zopiclone N-oxide, (5RS)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate 4-oxide, Zopiclone oxide, Zopiclone Imp. A (EP)
IUPAC-namn [6-(5-chloropyridin-2-yl)-5-oxo-7H-pyrrolo[3,4-b]pyrazin-7-yl] 4-methyl-4-oxidopiperazin-4-ium-1-carboxylate
LEDER C[N+]1([O-])CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4
Specifikationer
Kemiskt namn eller material Zopiclone-N-oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C17H17ClN6O4/c1-24(27)8-6-22(7-9-24)17(26)28-16-14-13(19-4-5-20-14)15(25)23(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3
Formel vikt 404.1
Procent renhet >95%