missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZWINT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12282-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
ZWINT | |
Polyclonal | |
Unconjugated | |
ZWINT | |
HZwint 1, KNTC2AP, ZW10 interacting protein 1, ZW10 interactor, ZWINT, Zwint 1, ZWINT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11130 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95229 | |
ZWINT | |
ZWINT Fusion Protein Ag2933 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |