Läs mer
Zymostenol, TRC
Produktkod. 30453354
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453354 10 mg
30453118 50 mg
30455673 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30453354 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ70153010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 566-97-2
Molekylformel C27 H46 O
Molekylvikt (g/mol) 386.65
Synonym 5α-Cholest-8-en-3β-ol, 3β-Hydroxy-5α-cholest-8(9)-ene, Dihydrozymosterol, δ8-Cholestenol, (3β,5α)-Cholest-8-en-3-ol
IUPAC-namn (3S,5S,10S,13R,14R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](O)C[C@@H]4CC3
Specifikationer
Kemiskt namn eller material Zymostenol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-21,23-24,28H,6-17H2,1-5H3/t19-,20+,21+,23-,24+,26+,27-/m1/s1
Formel vikt 386.3549
Procent renhet >95%