missing translation for 'onlineSavingsMsg'
Learn More

Zyxin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10330-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826653

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Zyxin binds alpha-actinin and the CRP proteins and is implicated in the regulation and modulation of actin filaments organization, cell adhesion, cell motility, mitosis, and signal transduction. Zyxin may shuttle between focal adhesion plaques and the nucleus. It has been suggested that Zyxin is involved in gene transcription by its association with transcription factors in the nucleus.
Specifications

Specifications

Zyxin
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15942
ESP 2, HED 2, ZYX, zyxin, Zyxin 2
Rabbit
IgG
20 μL
-20°C
Primary
7791
Western Blot
Unconjugated
ZYX
Liquid
ZYX
Zyxin Fusion Protein Ag0419
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers