CAS RN 1034439-43-4
CAS RN 1034439-43-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
5,6-Dimethoxy-2-[(4-piperidyl)methyl]indane Hydrochloride(Donepezil Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,6-Dimethoxy-2-[(4-piperidyl)methyl]indane Hydrochloride(Donepezil Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.