CAS RN 1034442-47-1
CAS RN 1034442-47-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
8-Isothiocyanato-1,3,6-pyrenetrisulfonic Acid Trisodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
8-Isothiocyanato-1,3,6-pyrenetrisulfonic Acid Trisodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.