CAS RN 1034706-81-4
CAS RN 1034706-81-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(+)-N-Acetyl 3,4,4a,5,6,10b-Hexahydro-2H-naphtho[1,2-b][1,4]oxazine-9-ol Triisopropylsilyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-N-Acetyl 3,4,4a,5,6,10b-Hexahydro-2H-naphtho[1,2-b][1,4]oxazine-9-ol Triisopropylsilyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(+)-N-Acetyl 3,4,4a,5,6,10b-Hexahydro-2H-naphtho[1,2-b][1,4]oxazine-9-ol Triisopropylsilyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.