CAS RN 103929-85-7
CAS RN 103929-85-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
1-(Chloromethyl)-3,5-bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(Chloromethyl)-3,5-bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.