CAS RN 1041-01-6
CAS RN 1041-01-6
IUPAC Namn:
2-amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid
Synonymer:
3,5-diiodothyronine 3,5-diiodo-dl-thryronine
Molekylvikt (g/mol):
525.08
Molekylär sammansättning:
C15H13I2NO4
InChi Key:
ZHSOTLOTTDYIIK-UHFFFAOYNA-N
SMILES:
NC(CC1=CC(I)=C(OC2=CC=C(O)C=C2)C(I)=C1)C(O)=O
1
–
1
av
1
Resultat
1
(2S)-2-Amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic Acid (Diiodothyronine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.