CAS RN 104758-14-7
CAS RN 104758-14-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(6R,7R)-3-(Acetoxymethyl)-8-oxo-7-(thiophene-2-carboxamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(6R,7R)-3-(Acetoxymethyl)-8-oxo-7-(thiophene-2-carboxamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(6R,7R)-3-(Acetoxymethyl)-8-oxo-7-(thiophene-2-carboxamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.