CAS RN 10487-71-5
CAS RN 10487-71-5
IUPAC Namn:
(2E)-but-2-enoyl chloride
Synonymer:
2-butenoyl chloride
Molekylvikt (g/mol):
104.53
Molekylär sammansättning:
C4H5ClO
InChi Key:
RJUIDDKTATZJFE-NSCUHMNNSA-N
SMILES:
C\C=C\C(Cl)=O
1
–
2
av
2
Resultat
1
rac-Crotonoyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Crotonoyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.