CAS RN 105-21-5
CAS RN 105-21-5
IUPAC Namn:
5-propyloxolan-2-one
Synonymer:
gamma-heptalactone
Molekylvikt (g/mol):
128.17
Molekylär sammansättning:
C7H12O2
InChi Key:
VLSVVMPLPMNWBH-UHFFFAOYNA-N
SMILES:
CCCC1CCC(=O)O1
1
–
3
av
3
Resultat
1
gamma-Heptalactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Gamma-Heptalactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
gamma-Heptalactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.