CAS RN 10538-64-4
CAS RN 10538-64-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(3Alpha,5Beta,7Alpha)-3,7-Dihydroxy-12-oxocholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3a,5b,7a)-3,7-Dihydroxy-12-oxocholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3Alpha,5Beta,7Alpha)-3,7-Dihydroxy-12-oxocholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.