CAS RN 10548-46-6
CAS RN 10548-46-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
1,6-Anhydro-2,3,4-tri-O-benzyl-Beta-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,6-Anhydro-2,3,4-tri-O-benzyl-Beta-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,6-Anhydro-2,3,4-tri-O-benzyl-Beta-D-glucopyranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.