CAS RN 105807-84-9
CAS RN 105807-84-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
6-Amino-2,2-dimethyl-2H-benzo[b][1,4]oxazin-3(4h)one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Amino-2,2-dimethyl-2H-benzo[b][1,4]oxazin-3(4h)one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.