CAS RN 106-73-0
CAS RN 106-73-0
IUPAC Namn:
methyl heptanoate
Synonymer:
methyl heptanoate
Molekylvikt (g/mol):
144.21
Molekylär sammansättning:
C8H16O2
InChi Key:
XNCNNDVCAUWAIT-UHFFFAOYSA-N
SMILES:
CCCCCCC(=O)OC
1
–
2
av
2
Resultat
1
Methyl Heptanoate (Heptanoic Acid Methyl Ester), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl Heptanoate(Heptanoic Acid Methyl Ester), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.