CAS RN 1070-87-7
CAS RN 1070-87-7
IUPAC Name:
2,2,4,4-tetramethylpentane
Synonyms:
2,2,4,4-tetramethylpentane
Molecular Weight (g/mol):
128.26
Molecular Formula:
C9H20
InChi Key:
GUMULFRCHLJNDY-UHFFFAOYSA-N
SMILES:
CC(C)(C)CC(C)(C)C
1
–
1
of
1
results
1
2,2,4,4-Tetramethylpentane, 98%, Thermo Scientific Chemicals
CAS: 1070-87-7 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.26 MDL Number: MFCD00060873 InChI Key: GUMULFRCHLJNDY-UHFFFAOYSA-N PubChem CID: 14058 IUPAC Name: 2,2,4,4-tetramethylpentane SMILES: CC(C)(C)CC(C)(C)C