accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1072944-23-0

CAS RN 1072944-23-0

IUPAC Name: (6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
Synonyms: 6-bromo-2,4-dimethylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 229.87
Molecular Formula: C7H9BBrNO2
InChi Key: GXXITYQLWOMIEM-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=NC(C)=C1B(O)O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products