accessibility menu, dialog, popup

UserName

CAS RN 107333-47-1

CH3CH3CH3CH3CH3CH3CH3CH3CTi

CAS RN 107333-47-1

IUPAC Name: titanium(4+) cyclopenta-1,3-dien-1-ide octamethanide
Synonyms: titanium(4+) cyclopenta-1,3-dien-1-ide octamethanide
Molecular Weight (g/mol): 233.25
Molecular Formula: C13H29Ti
InChi Key: DVHMCUKDWMNRGX-UHFFFAOYSA-N
SMILES: [CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[Ti+4].C1C=CC=[C-]1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results