CAS RN 1076-96-6
CAS RN 1076-96-6
IUPAC Name:
methyl 4-ethenylbenzoate
Synonyms:
methyl 4-ethenylbenzoate
Molecular Weight (g/mol):
162.19
Molecular Formula:
C10H10O2
InChi Key:
NUMHUJZXKZKUBN-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C)C=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Methyl 4-vinylbenzoate, 96%
CAS: 1076-96-6 Molecular Formula: C10H10O2 Molecular Weight (g/mol): 162.19 MDL Number: MFCD00017218 InChI Key: NUMHUJZXKZKUBN-UHFFFAOYSA-N Synonym: methyl 4-vinylbenzoate,methyl4-vinylbenzoate,methyl p-vinylbenzoate,methyl-4-vinylbenzoate,benzoic acid, 4-ethenyl-, methyl ester,benzoic acid, p-vinyl-, methyl ester,4-vinyl benzoic acid methyl ester,acmc-1brgo,4-vinyl-benzoic acid methyl ester PubChem CID: 583124 IUPAC Name: methyl 4-ethenylbenzoate SMILES: COC(=O)C1=CC=C(C=C)C=C1